missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclohexanediacetic anhydride, Thermo Scientific™

Product Code. 17780629 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17780629 5 g 5g
17790629 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17780629 Supplier Thermo Scientific Acros Supplier No. 358130050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Alpha Vector CYCLOHEXANEDIACETICANHYDRIDE
Chemical Name or Material 1,1-Cyclohexanediacetic anhydride
CAS 1010-26-0
Melting Point 67°C to 72°C
Color Orange to White
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C10 H14 O3
MDL Number 00040292
Quantity 5 g
Synonym 3-Oxaspiro[5.5 ]undecane-2,4-dione
Molecular Weight (g/mol) 182.22
Physical Form Crystalline Powder or Crystals
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.