missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclohexanediacetic acid, 98%

Product Code. 11425746
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425746 50 g 50g
11415746 10 g 10g
11435746 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425746 Supplier Thermo Scientific Alfa Aesar Supplier No. B20816.18
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4355-11-7 | C10H16O4 | 200.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4355-11-7
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00001519
InChI Key YQPCHPBGAALCRT-UHFFFAOYSA-N
Synonym 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
PubChem CID 78061
IUPAC Name 2-[1-(carboxymethyl)cyclohexyl]acetic acid
SMILES C1CCC(CC1)(CC(=O)O)CC(=O)O

Specifications

Melting Point 178°C to 182°C
Quantity 50 g
Formula Weight 200.23
Percent Purity 98%
Chemical Name or Material 1,1-Cyclohexanediacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.