missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Chloro-5-isoquinolinesulfonyl)homopiperazine, TRC

Product Code. 30368144
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368144 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368144 Supplier Toronto Research Chemicals Supplier No. TRCC36765050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 105628-70-4
Molecular Formula C14 H16 Cl N3 O2 S
Quantity 50 mg
Synonym Fasudil Impurity 33 HCl
InChI Formula InChI=1S/C14H16ClN3O2S/c15-14-12-3-1-4-13(11(12)5-7-17-14)21(19,20)18-9-2-6-16-8-10-18/h1,3-5,7,16H,2,6,8-10H2
SMILES Clc1nccc2c(cccc12)S(=O)(=O)N3CCCNCC3
IUPAC Name 1-chloro-5-(1,4-diazepan-1-ylsulfonyl)isoquinoline
Molecular Weight (g/mol) 325.81
Formula Weight 325.0652
Chemical Name or Material 1-(1-Chloro-5-isoquinolinesulfonyl)homopiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.