missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Carbonyldipyrrolidine, 98%, Thermo Scientific™

Product Code. 15417448
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15417448

Brand: Thermo Scientific Alfa Aesar H60942.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Reactant in the ring-opening polymerization of six-and seven-membered cyclic pseudoureas.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81759-25-3
Molecular Formula C9H16N2O
Molecular Weight (g/mol) 168.24
MDL Number MFCD00216641
InChI Key HPTQKSXAQBHFKL-UHFFFAOYSA-N
Synonym 1,1'-carbonyldipyrrolidine, di pyrrolidin-1-yl methanone, 1,1-carbonyldipyrrolidine, 1-pyrrolidine-1-carbonyl pyrrolidine, pyrrolidine, 1,1'-carbonylbis, ditetrahydro-1h-pyrrol-1-ylmethanone, carbodipiperidide, dipyrrolizinoketone, carbonyldipyrrolidine, dipyrrolidinyl ketone
PubChem CID 736440
IUPAC Name dipyrrolidin-1-ylmethanone
SMILES O=C(N1CCCC1)N1CCCC1

Specifications

Color Colorless to Yellow
Density 1.086 g/mL
Boiling Point 148°C to 150°C
Flash Point 113°C (235°F)
Refractive Index 1.513
Quantity 50 g
Beilstein 1637732
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 168.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,1'-Carbonyldipyrrolidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.