missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(hydroxymethyl)cyclopropane, 90%, Thermo Scientific™

Product Code. 10031084 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031084 5 g Glass bottle 5g
10547454 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10031084 Supplier Thermo Scientific Acros Supplier No. 426250050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 39590-81-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00464207
InChI Key YAINYZJQSQEGND-UHFFFAOYSA-N
Synonym 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f
PubChem CID 4101437
IUPAC Name [1-(hydroxymethyl)cyclopropyl]methanol
SMILES C1CC1(CO)CO

Specifications

CAS Min % 88.0
CAS Max % 100.0
Density 1.0650g/mL
Boiling Point 215.0°C to 233.0°C
Flash Point 110°C
Assay Percent Range 90%
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.065
Formula Weight 102.13
Percent Purity 90%
Chemical Name or Material 1, 1-Bis(hydroxymethyl)cyclopropane
Show More Show Less
compliance-icons
Product Identifier
  • 1,1-Bis(hydroxymethyl)cyclopropane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.