Learn More
1,1'-Bis(diphenylphosphino)ferrocene, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 12150-46-8 |
---|---|
Molecular Formula | C34H28FeP2 |
Molecular Weight (g/mol) | 554.39 |
MDL Number | MFCD00001422 |
InChI Key | KZPYGQFFRCFCPP-UHFFFAOYSA-N |
IUPAC Name | λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide) |
SMILES | [Fe].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12150-46-8 | |
554.39 | |
KZPYGQFFRCFCPP-UHFFFAOYSA-N | |
[Fe].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C34H28FeP2 | |
MFCD00001422 | |
λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide) |
Specifications
12150-46-8 | |
100.0 | |
Authentic | |
C34H28FeP2 | |
15, 3490 | |
[Fe].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
554.39 | |
≥97.5% | |
1, 1'-Bis(diphenylphosphino)ferrocene |
97.5 | |
Orange | |
98% | |
MFCD00001422 | |
KZPYGQFFRCFCPP-UHFFFAOYSA-N | |
λ²-iron(2+) bis(1-(diphenylphosphanyl)cyclopenta-2,4-dien-1-ide) | |
553.99 | |
Powder |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- 1,1``-Bis(diphenylphosphino)ferrocene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only