missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(diisopropylphosphino)ferrocene, 98%, Thermo Scientific Chemicals

Product Code. 11496100
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496100 1 g 1g
11406110 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11496100 Supplier Thermo Scientific Alfa Aesar Supplier No. H27057.03
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

The general and efficient cross-coupling of thiols with aryl halides was developed utilizing Pd(OAc) 2 /1,1-bis(diisopropylphosphino)ferrocene as the catalyst. The oxidative electrochemistry of dippf was examined, and it was found to follow an ECDim mechanism. Seven new compounds containing dippf were prepared and characterized spectroscopically. The crystal structures of [MCl2(dippf)] (M = Pt and Zn) were determined.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The general and efficient cross-coupling of thiols with aryl halides was developed utilizing Pd(OAc) 2 /1,1′-bis(diisopropylphosphino)ferrocene as the catalyst. The oxidative electrochemistry of dippf was examined, and it was found to follow an ECDim mechanism. Seven new compounds containing dippf were prepared and characterized spectroscopically. The crystal structures of [MCl2(dippf)] (M = Pt and Zn) were determined.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97239-80-0
Molecular Formula C22H36FeP2
Molecular Weight (g/mol) 418.32
MDL Number MFCD02684559
IUPAC Name 1,1'-Bis(diisopropylphosphino)ferrocene
SMILES [Fe].CC(C)P(C(C)C)c1cccc1.CC(C)P(C(C)C)c1cccc1

Specifications

Melting Point 50°C to 52°C
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 418.34
Percent Purity 98%
Chemical Name or Material 1,1'-Bis(diisopropylphosphino)ferrocene
compliance-icons
Product Identifier
  • 1,1'-Bis(diisopropylphosphino)ferrocene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,1-Bis(diisopropylphosphino)ferrocene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.