missing translation for 'onlineSavingsMsg'
Learn More

1,1’-Bicyclohexyl, TRC

Product Code. 30338623
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338623 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338623 Supplier Toronto Research Chemicals Supplier No. TRCB382880100ML
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dicyclohexyl
CAS 92-51-3
Purity Grade Notes GC
Molecular Formula C12 H22
Quantity 100 mL
Synonym Bicyclohexyl (6CI,8CI), 1,1'-Biphenyl, dodecahydro-, Cyclohexane, cyclohexyl-, Cyclohexylcyclohexane, Dicyclohexane, Dicyclohexyl, Dodecahydrobiphenyl, NSC 59855
InChI Formula InChI=1S/C12H22/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h11-12H,1-10H2
SMILES C1CCC(CC1)C2CCCCC2
IUPAC Name cyclohexylcyclohexane
Molecular Weight (g/mol) 166.3
Formula Weight 166.1722
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.