missing translation for 'onlineSavingsMsg'
Learn More

(±)-1,1'-Bi(2-naphthylamine), 97%

Product Code. 15454057
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15454057

Brand: Thermo Scientific Alfa Aesar H31264.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4488-22-6 | C20H16N2 | 284.362 g/mol

Novel axially chiral Rh N-heterocyclic carbene complexes were prepared from axially dissymmetric 1,1?-binaphthalenyl-2,2?-diamine and applied in the Rh-catalyzed enantioselective hydrosilylation of methyl ketones. The enantioselective direct aldol reaction, organocatalyzed by recoverable BINAM- prolinamide derivatives can be highly accelerated by a catalytic amount of a carboxylic acid without a detrimental of the obtained enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Novel axially chiral Rh N-heterocyclic carbene complexes were prepared from axially dissymmetric 1,1′-binaphthalenyl-2,2′-diamine and applied in the Rh-catalyzed enantioselective hydrosilylation of methyl ketones. The enantioselective direct aldol reaction, organocatalyzed by recoverable BINAM- prolinamide derivatives can be highly accelerated by a catalytic amount of a carboxylic acid without a detrimental of the obtained enantioselectivities.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4488-22-6
Molecular Formula C20H16N2
Molecular Weight (g/mol) 284.362
MDL Number MFCD00145204
InChI Key DDAPSNKEOHDLKB-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
PubChem CID 20571
IUPAC Name 1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N

Specifications

Melting Point 188°C to 193°C
Quantity 1 g
Solubility Information Insoluble in water
Formula Weight 284.36
Percent Purity 97%
Chemical Name or Material (±)-1,1'-Bi(2-naphthylamine)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.