missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1,1'-Bi(2-naphthol), 99%

Product Code. 11439123
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439123 5 g 5g
11449123 25 g 25g
11459123 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439123 Supplier Thermo Scientific Alfa Aesar Supplier No. A11182.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 602-09-5 | C20H14O2 | 286.33 g/mol

(±)-1,1'-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-1,1′-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

Solubility
Soluble in dioxane (50 mg/mL). Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Refrigerated (2-8°C)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 602-09-5
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00004068
InChI Key PPTXVXKCQZKFBN-UHFFFAOYSA-N
Synonym 1,1'-bi-2-naphthol, s---1,1'-bi-2-naphthol, r-+-1,1'-bi-2-naphthol, 1,1'-binaphthalene-2,2'-diol, binol, 1,1'-binaphthyl-2,2'-diol, 2,2'-dinaphthol, beta-binaphthol, r-1,1'-bi-2-naphthol, s-1,1'-binaphthalene-2,2'-diol
PubChem CID 11762
IUPAC Name 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O

Specifications

Melting Point 215°C to 218°C
Density 1.301
Odor Characteristic
Quantity 5 g
UN Number UN2811
Beilstein 997518
Merck Index 14,1226
Solubility Information Soluble in dioxane (50mg/mL). Insoluble in water.
Formula Weight 286.33
Percent Purity 99%
Chemical Name or Material (±)-1,1'-Bi (2-naphthol)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.