missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Benzothiophen-2-yl)-2-bromo-1-ethanone, ≥97%, Thermo Scientific™

Product Code. 10631085
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 10631085

Brand: Thermo Scientific Maybridge CC12205CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 97511-06-3
Molecular Formula C10H7BrOS
Molecular Weight (g/mol) 255.13
MDL Number MFCD06658971
InChI Key WVTSOGFICBVCRE-UHFFFAOYSA-N
Synonym 1-1-benzothiophen-2-yl-2-bromo-1-ethanone, 1-1-benzothiophen-2-yl-2-bromoethanone, 2-bromoacetyl benzo b thiophene, 1-1-benzothiophen-2-yl-2-bromoethan-1-one, 1-benzo b thiophen-2-yl-2-bromoethanone, 1-benzo b thiophen-2-yl-2-bromoethan-1-one, 1-benzothiophen-2-yl-2-bromoethan-1-one, 2-bromoacetyl thianaphthene
PubChem CID 2795258
IUPAC Name 1-(1-benzothiophen-2-yl)-2-bromoethanone
SMILES BrCC(=O)C1=CC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-(1-Benzothiophen-2-yl)-2-bromo-1-ethanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.