missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone, 95+%, Thermo Scientific™

Product Code. 10681863
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10681863

Brand: Thermo Scientific Maybridge CC27505CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 187657-92-7
Molecular Formula C10H7BrO2
Molecular Weight (g/mol) 239.07
MDL Number MFCD03407369
InChI Key WVHFTONHSRLBGL-UHFFFAOYSA-N
Synonym 1-benzofuran-3-yl-2-bromoethanone, 1-1-benzofuran-3-yl-2-bromo-1-ethanone, 1-1-benzofuran-3-yl-2-bromoethanone, 1-benzofuran-3-yl-2-bromoethan-1-one, 1-1-benzofuran-3-yl-2-bromoethan-1-one, ethanone,1-3-benzofuranyl-2-bromo, 1-benzofuran-3-yl-2-bromo-1-ethanone, pubchem7014, 3-2-bromoacetyl benzo b furan, 1-benzofuran-3-yl-2-bromo-ethanone
PubChem CID 2776580
IUPAC Name 1-(1-benzofuran-3-yl)-2-bromoethanone
SMILES BrCC(=O)C1=COC2=CC=CC=C12

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 239.07
Percent Purity 95%
Chemical Name or Material 1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.