missing translation for 'onlineSavingsMsg'
Learn More

1,1'-(Azodicarbonyl)dipiperidine, TRC

Product Code. 30340334
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30340334

Brand: Toronto Research Chemicals TRCA965410500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1'-(Azodicarbonyl)dipiperidine
CAS 10465-81-3
Molecular Formula C12H20N4O2
Quantity 500 mg
InChI Formula InChI=1S/C12H20N4O2/c17-11(15-7-3-1-4-8-15)13-14-12(18)16-9-5-2-6-10-16/h1-10H2
SMILES O=C(N=NC(=O)N1CCCCC1)N2CCCCC2
IUPAC Name N-(piperidine-1-carbonylimino)piperidine-1-carboxamide
Molecular Weight (g/mol) 252.31
Formula Weight 252.1586
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.