missing translation for 'onlineSavingsMsg'
Learn More

1,1'-(Azodicarbonyl)dipiperidine, 97%

Product Code. 11382839
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382839 5 g 5g
11392839 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382839 Supplier Thermo Scientific Alfa Aesar Supplier No. H27719.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10465-81-3 | C12H20N4O2 | 252.32 g/mol

It is used in a study of the copper-catalyzed addition of aryboronic acids to azodicarboxyl derivatives providing aryl-substituted hydrazides. It is widely used reagent for the Mitsunobu reaction. It is reactant for preparation of polyfluoroalkylated tripyrazolylmethane ligands, (-)-Hygromycin A via Mitsunobu glycosylation, optically active α,α-disubstituted amino acids via Mitsunobu reaction, aza-β-lactams through [2+2] cycloaddition reactions, glycosyl disulfides, pyridine ether PPAR agonists, S-glycosyl amino acid, building blocks for combinatorial neoglycopeptide synthesis and is a histamine H3 receptor antagonists. It is a reactant for Mitsunobu inversion reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in a study of the copper-catalyzed addition of aryboronic acids to azodicarboxyl derivatives providing aryl-substituted hydrazides. It is widely used reagent for the Mitsunobu reaction. It is reactant for preparation of polyfluoroalkylated tripyrazolylmethane ligands, (-)-Hygromycin A via Mitsunobu glycosylation, optically active α,α-disubstituted amino acids via Mitsunobu reaction, aza-β-lactams through [2+2] cycloaddition reactions, glycosyl disulfides, pyridine ether PPAR agonists, S-glycosyl amino acid, building blocks for combinatorial neoglycopeptide synthesis and is a histamine H3 receptor antagonists. It is a reactant for Mitsunobu inversion reactions.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10465-81-3
Molecular Formula C12H20N4O2
Molecular Weight (g/mol) 252.32
MDL Number MFCD00010111
InChI Key OQJBFFCUFALWQL-UHFFFAOYSA-N
Synonym 1,1'-azodicarbonyl-dipiperidine, 1,1-azodicarbonyl dipiperidine, 1,1'-azodicarbonyl dipiperidine, addp, diazene-1,2-diylbis piperidin-1-ylmethanone, azodicarboxylic acid dipiperidine, ne-n-piperidine-1-carbonylimino piperidine-1-carboxamide, 1,1'-azodicarbonyl dipiperidine addp, piperidyl piperidylcarbonyl diazenyl ketone, e-diazene-1,2-diylbis piperidin-1-ylmethanone
PubChem CID 5702657
IUPAC Name (NE)-N-(piperidine-1-carbonylimino)piperidine-1-carboxamide
SMILES O=C(N=NC(=O)N1CCCCC1)N1CCCCC1

Specifications

Melting Point 133°C to 136°C
Odor Odorless
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 252.32
Percent Purity 97%
Chemical Name or Material 1,1'-(Azodicarbonyl)dipiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.