missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Azobis(N,N-dimethylformamide), 95%

Product Code. 15426619
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426619 1 g 1g
15436619 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426619 Supplier Thermo Scientific Alfa Aesar Supplier No. J64865.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10465-78-8 | C6H12N4O2 | 172.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.188
MDL Number MFCD00008318
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C

Specifications

Melting Point 112°C
Color Yellow
Linear Formula (CH3)2NCON=NCON(CH3)2
Quantity 1 g
Beilstein 1910409
Formula Weight 172.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1,1'-Azobis(N,N-dimethylformamide)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.