missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Adamantyl)ethylamine hydrochloride, Thermo Scientific™

Product Code. p-2693951
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10633203

missing translation for 'mfr': Thermo Scientific Maybridge BTB12981DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1501-84-4
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00072023
InChI Key OZBDFBJXRJWNAV-UHFFFAOYNA-N
Synonym rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
PubChem CID 15165
ChEBI CHEBI:8865
IUPAC Name 1-(1-adamantyl)ethanamine;hydrochloride
SMILES [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.77
Percent Purity 95%
Chemical Name or Material 1-(1-Adamantyl)ethylamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.