missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Adamantyl)ethylamine hydrochloride, 98%

Product Code. 15420138
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420138 25 g 25g
15410138 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420138 Supplier Thermo Scientific Alfa Aesar Supplier No. H53377.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1501-84-4 | C12H22ClN | 215.77 g/mol

1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).

Solubility
Freely soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1501-84-4
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00072023
InChI Key OZBDFBJXRJWNAV-UHFFFAOYNA-N
Synonym rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
PubChem CID 15165
ChEBI CHEBI:8865
IUPAC Name 1-(1-adamantyl)ethanamine;hydrochloride
SMILES [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 375°C
Quantity 25 g
Merck Index 148224
Solubility Information Freely soluble in water.
Formula Weight 215.77
Percent Purity 98%
Chemical Name or Material 1-(1-Adamantyl)ethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.