missing translation for 'onlineSavingsMsg'
Learn More

1-[1-(4-Fluorobenzyl)-2-benzimidazolyl]-4-(methylamino)piperidine dihydrochloride, 97%, Thermo Scientific™

Product Code. 15465645
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15465645

Brand: Thermo Scientific Alfa Aesar H63927.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1134322-93-2
Molecular Formula C20H25Cl2FN4
MDL Number MFCD03844681
Quantity 250 mg
Synonym 1-1-4-fluorophenyl methyl-1h-benzimidazol-2-yl-n-methyl-4-piperidinamine dihydrochloride, 1-1-4-fluorophenyl methyl-1h-benzimidazol-2-yl-4-methylaminopiperidine dihydrochloride, 1-1-4-fluorophenyl methyl-1,3-benzodiazol-2-yl-n-methylpiperidin-4-amine dihydrochloride
InChI Key ZIGFBTWAPHHQRA-UHFFFAOYSA-N
SMILES CNC1CCN(CC1)C2=NC3=CC=CC=C3N2CC4=CC=C(C=C4)F.Cl.Cl
IUPAC Name 1-[1-[(4-fluorophenyl)methyl]benzimidazol-2-yl]-N-methylpiperidin-4-amine;dihydrochloride
Molecular Weight (g/mol) 411.346
PubChem CID 24720879
Formula Weight 411.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-[1-(4-Fluorobenzyl)-2-benzimidazolyl]-4-(methylamino)piperidine dihydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.