missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,7,7-Pentamethyldiethylenetriamine, TRC

Product Code. 30417831
Click to view available options
Quantity:
10 mg
1 mL
Unit Size:
10mg
1mL
This item is not returnable. View return policy

Product Code. 30417831

Brand: Toronto Research Chemicals TRCP22747010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentamethyldiethylenetriamine
CAS 3030-47-5
Molecular Formula C9 H23 N3
Quantity 10 mg
Synonym 1,2-Ethanediamine','N-[2-(dimethylamino)ethyl]-N,N',N'-trimethyl- (9CI), Diethylenetriamine','1,1,4,7,7-pentamethyl- (6CI,7CI,8CI), N1-[2-(Dimethylamino)ethyl]-N1,N2,N2-trimethyl-1,2-ethanediamine, (2-[[2-(Dimethylamino)ethyl](methyl)amino]ethyl)dimethylamine, 1,1,4,7,7-Pentamethyl-1,4,7-triazaheptane, 1,1,4,7,7-Pentamethyldiethylenetriamine, 2,5,8-Trimethyl-2,5,8-triazanonane, AM 1, Bis[2-(dimethylamino)ethyl]methylamine, DY 5, Dabco PMDETA, Desmorapid PV, Jeffcat PMDETA, KL 3, Kao 3, Kaolizer 3, Lupragen N 301, N 301, N,N,N',N',N''-Pentamethylbis(2-aminoethyl)amine, N,N,N',N'',N''-Pentamethyl-3-azapentane-1,5-diamine, N,N,N',N'',N''-Pentamethylbis(2-aminoethyl)amine, N,N,N',N'',N''-Pentamethyldiethylenetriamine, N,N,N',N'',N''-Pentamethyldiethylenetriamine, N,N-Bis(2-dimethylaminoethyl)methanamine, N,N-Bis[2-(dimethylamino)ethyl]methylamine, N-Methyl-N,N-bis(2-dimethylaminoethyl)amine, N-[2-(Dimethylamino)ethyl]-N,N',N'-trimethyl-1,2-ethanediamine, N1,N1,N3,N5,N5-Pentamethyl-3-azapentane-1,5-diamine, NSC 65659, Niax C 5, Niax Catalyst C 5, PC 5, PC CAT PMDETA, PMDETA, PMDT, PMDTA, Pentamethyldiethylenetriamine, Polycat 5, Tegoamin PMDETA, Toyocat DT
InChI Formula InChI=1S/C9H23N3/c1-10(2)6-8-12(5)9-7-11(3)4/h6-9H2,1-5H3
SMILES CN(C)CCN(C)CCN(C)C
IUPAC Name N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine
Molecular Weight (g/mol) 173.299
Formula Weight 173.1892
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.