missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,7,7-Pentamethyldiethylenetriamine, 98%

Product Code. 11418426
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418426 25 g 25g
11428426 100 g 100g
11438426 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418426 Supplier Thermo Scientific Alfa Aesar Supplier No. A15433.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3030-47-5 | C9H23N3 | 173.304 g/mol

Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

Solubility
Miscible in water.

Notes
Air & Moisture Sensitive. Store under inert gas. Incompatible with strong oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3030-47-5
Molecular Formula C9H23N3
Molecular Weight (g/mol) 173.304
MDL Number MFCD00014876
InChI Key UKODFQOELJFMII-UHFFFAOYSA-N
Synonym pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine
PubChem CID 18196
ChEBI CHEBI:39475
IUPAC Name N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine
SMILES CN(C)CCN(C)CCN(C)C

Specifications

Melting Point <-20°C
Density 0.83
Boiling Point 199°C
Flash Point 72°C (161°F)
Refractive Index 1.442
Quantity 25 g
UN Number UN2922
Beilstein 1741396
Sensitivity Air and moisture sensitive
Solubility Information Miscible in water.
Formula Weight 173.3
Percent Purity 98%
Chemical Name or Material 1,1,4,7,7-Pentamethyldiethylenetriamine
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.