missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%

Product Code. 11493447
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493447 1 g 1g
11403457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493447 Supplier Thermo Scientific Alfa Aesar Supplier No. A17225.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1450-63-1 | C28H22 | 358.48 g/mol

1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.

Solubility
Soluble in ethanol, benzene, chloroform, tolune and acetic acid.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-63-1
Molecular Formula C28H22
Molecular Weight (g/mol) 358.48
MDL Number MFCD00004766
InChI Key KLCLIOISYBHYDZ-UHFFFAOYSA-N
Synonym 1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene
PubChem CID 74060
SMILES C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 197°C to 202°C
Quantity 1 g
Solubility Information Soluble in ethanol,benzene,chloroform,tolune and acetic acid.
IUPAC Name 1,4,4-triphenylbuta-1,3-dienylbenzene
Formula Weight 358.48
Percent Purity 99%
Chemical Name or Material 1,1,4,4-Tetraphenyl-1,3-butadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.