missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylurea, 99%

Product Code. 10103430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103430 250 mL Glass bottle 250mL
10386920 100 mL Glass bottle 100mL
10637943 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103430 Supplier Thermo Scientific Acros Supplier No. 138492500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 632-22-4 | C5H12N2O | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-22-4
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008319
InChI Key AVQQQNCBBIEMEU-UHFFFAOYSA-N
Synonym tetramethylurea, temur, urea, tetramethyl, n,n,n',n'-tetramethylurea, tetramethyluree, tmu, urea, 1,1,3,3-tetramethyl, urea, n,n,n',n'-tetramethyl, tetramethyluree french, tetramethyl-urea
PubChem CID 12437
ChEBI CHEBI:84278
IUPAC Name 1,1,3,3-tetramethylurea
SMILES CN(C)C(=O)N(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -1.0°C
Color Colorless to Yellow
Density 0.9879g/mL
Boiling Point 174.0°C to 178.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCON(CH3)2
Refractive Index 1.4496 to 1.4516
Quantity 250 mL
Beilstein 04, 74
Fieser 01,1146; 13,302; 15,305
Merck Index 15, 9378
Specific Gravity 0.9879
Solubility Information Solubility in water: 116g/L. Other solubilities: miscible with all common organic solvents,,including petroleum ether
Viscosity 1.5 mPa.s (20°C)
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethylurea, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,3,3-Tetramethylurea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tetramethylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.