missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyldisiloxane, 97%, Thermo Scientific Chemicals

Product Code. 10711301
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3277-26-7
Molecular Formula C4H14OSi2
Molecular Weight (g/mol) 134.33
InChI Key KWEKXPWNFQBJAY-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010
PubChem CID 6327482
IUPAC Name dimethylsilylideneoxonio(dimethyl)silanide
SMILES C[Si-](C)[O+]=[Si](C)C

Specifications

Boiling Point 70.0°C to 71.0°C
Melting Point -78.0°C
Color Colorless
Physical Form Liquid
Quantity 25 mL
Assay Percent Range 97%
Linear Formula [(CH3)2SiH]2O
Flash Point −20°C
Solubility Information Solubility in water: decomposes. Other solubilities: decomposes with alcohol
Formula Weight 134.33
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3690 to 1.3710
Specific Gravity 0.76
Density 0.7600g/mL
Chemical Name or Material 1,1,3,3-Tetramethyldisiloxane
Show More Show Less
compliance-icons
Product Identifier
  • 1,1,3,3-Tetramethyldisiloxane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'