missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyldisiloxane, 97%

Product Code. 10175030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175030 100 mL Glass bottle 100mL
10711301 25 mL Glass bottle 25mL
11965821 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175030 Supplier Thermo Scientific Acros Supplier No. 216511000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3277-26-7 | C4H14OSi2 | 134.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3277-26-7
Molecular Formula C4H14OSi2
Molecular Weight (g/mol) 134.33
InChI Key KWEKXPWNFQBJAY-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010
PubChem CID 6327482
IUPAC Name dimethylsilylideneoxonio(dimethyl)silanide
SMILES C[Si-](C)[O+]=[Si](C)C

Specifications

Boiling Point 70.0°C to 71.0°C
Melting Point -78.0°C
Color Colorless
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 97%
Linear Formula [(CH3)2SiH]2O
Flash Point −20°C
Solubility Information Solubility in water: decomposes. Other solubilities: decomposes with alcohol
Formula Weight 134.33
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3690 to 1.3710
Specific Gravity 0.76
Density 0.7600g/mL
Chemical Name or Material 1,1,3,3-Tetramethyldisiloxane
Show More Show Less
compliance-icons
Product Identifier
  • 1,1,3,3-Tetramethyldisiloxane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.