missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyl-1,3-divinyldisilazane, 96%, Thermo Scientific™

Product Code. 11339185
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339185 5 g 5g
11349185 25 g 25g
11359185 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339185 Supplier Thermo Scientific Alfa Aesar Supplier No. L16480.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,1,3,3-Tetramethyl-1,3-divinyldisilazane is used for silicone rubber, silicone resin, silicone gel and vinyl manufacturing.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7691-02-3
MDL Number MFCD00042846
IUPAC Name [[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene

Specifications

Boiling Point 160°C to 161°C
Quantity 5 g
Molecular Formula C8H19NSi2
UN Number UN2924
Beilstein 2411661
Flash Point 42°C (107°F)
Synonym 1,1,3,3-tetramethyl-1,3-divinyldisilazane, 1,3-divinyltetramethyldisilazane, 1,3-divinyl-1,1,3,3-tetramethyldisilazane, silanamine, 1-ethenyl-n-ethenyldimethylsilyl-1,1-dimethyl, bis dimethyl vinyl silyl amine, ccris 2450, bis ethenyldimethylsilyl amine, ethenyl-dimethylsilyl amino-dimethylsilyl ethene, n-dimethylvinylsilyl-1,1-dimethyl-1-vinylsilylamine, dvtmds
Solubility Information Hydrolyzes in water.
InChI Key WYUIWUCVZCRTRH-UHFFFAOYSA-N
SMILES C[Si](C)(C=C)N[Si](C)(C)C=C
Molecular Weight (g/mol) 185.42
PubChem CID 82126
Formula Weight 185.42
Percent Purity 96%
Refractive Index 1.44
Sensitivity Moisture sensitive
Density 0.819
Chemical Name or Material 1,1,3,3-Tetramethyl-1,3-divinyldisilazane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.