missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethoxypropane, 98%

Product Code. 11480253
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480253 1000 g 1000g
11490253 250 g 250g
11400263 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480253 Supplier Thermo Scientific Alfa Aesar Supplier No. A11418.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-52-3 | C7H16O4 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-52-3
Molecular Formula C7H16O4
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008488
InChI Key XHTYQFMRBQUCPX-UHFFFAOYSA-N
Synonym propane, 1,1,3,3-tetramethoxy, tetramethoxypropane, malonaldehyde bis dimethyl acetal, malonaldehyde tetramethyl acetal, malonaldehyde, bis dimethyl acetal, 1,1,3,3-tetramethoxy-propane, malonaldehyde bis dime, 1,3,3-tetramethoxypropane, ksc174o0b, propane,1,3,3-tetramethoxy
PubChem CID 66019
IUPAC Name 1,1,3,3-tetramethoxypropane
SMILES COC(CC(OC)OC)OC

Specifications

Density 0.997
Boiling Point 182°C to 183°C
Flash Point 54°C (129°F)
Refractive Index 1.407
Quantity 1000 g
UN Number UN3271
Beilstein 1700020
Sensitivity Moisture sensitive
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material 1,1,3,3-Tetramethoxypropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.