missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetraethoxypropane, TRC

Product Code. 30426376
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Unit Size:
1mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426376 25 mL 25mL
30414472 5 mL -
30410159 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426376 Supplier Toronto Research Chemicals Supplier No. TRCT29295725ML
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-31-6
Molecular Formula C11H24O4
Molecular Weight (g/mol) 220.31
Synonym Malonaldehyde Bis(diethyl Acetal), 1,1,3,3-Tetrakis(ethoxy)propane, 1,1,3,3-Triethoxypropane, Malonaldehyde Tetraethyl Acetal, Malonaldehyde Tetraethyl Diacetal, Malondialdehyde Tetraethyl Acetal, NSC 17068, TEP, Tetraethyl Malondialdehyde Acetal
IUPAC Name 1,1,3,3-tetraethoxypropane
SMILES CCOC(CC(OCC)OCC)OCC

Specifications

Quantity 25 mL
InChI Formula InChI=1S/C11H24O4/c1-5-12-10(13-6-2)9-11(14-7-3)15-8-4/h10-11H,5-9H2,1-4H3
Formula Weight 220.1675
Chemical Name or Material 1,1,3,3-Tetraethoxypropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.