missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetraethoxypropane-1,3-d2, TRC

Product Code. 30424712
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424712 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30424712 Supplier Toronto Research Chemicals Supplier No. TRCT29295550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1,3,3-Tetraethoxypropane-1,3-d2
CAS 105479-86-5
Purity Grade Notes GC
Molecular Formula C11 2H2 H22 O4
Quantity 50 mg
Synonym Propane-1,3-d2','1,1,3,3-tetraethoxy- (9CI), Malonaldehyde-1,3-d2 Bis(diethyl Acetal), 1,1,3,3-Tetraethoxypropane-1,3-d2, 1,1,3,3-Tetraethoxypropane-1,3-D2
InChI Formula InChI=1S/C11H24O4/c1-5-12-10(13-6-2)9-11(14-7-3)15-8-4/h10-11H,5-9H2,1-4H3/i10D,11D
SMILES [2H]C(CC([2H])(OCC)OCC)(OCC)OCC
IUPAC Name 1,3-dideuterio-1,1,3,3-tetraethoxypropane
Molecular Weight (g/mol) 222.32
Formula Weight 222.18
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.