missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5-Hexamethyltrisiloxane, 97%, ACROS Organics™

Product Code. 10737402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10737402 5mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10737402 Supplier Thermo Scientific Acros Supplier No. 337570050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 1189-93-1
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8220g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H18O2Si3
Linear Formula Si(CH3)2[OSi(CH3)2H]
Refractive Index 1.3790 to 1.3820 (20°C, 589nm)
MDL Number MFCD00039788
Quantity 5 mL
Synonym 1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,1,3,3,5,5-hexamethyl, 1,3,3,5,5-hexamethyltrisiloxane, 2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, dimethylbis dimethylsilyloxy silane
InChI Key YTEISYFNYGDBRV-UHFFFAOYSA-N
SMILES C[Si](C)O[Si](C)(C)O[Si](C)C
IUPAC Name [(dimethyl-$l^{3}-silanyl)oxy-dimethylsilyl]oxy-dimethylsilicon
Molecular Weight (g/mol) 206.463
PubChem CID 6327152
Formula Weight 208.48
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,1,3,3,5,5-Hexamethyltrisiloxane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.