missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5-Hexamethyltrisiloxane, 95%

Product Code. 11349215
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349215 100 g 100g
11339215 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349215 Supplier Thermo Scientific Alfa Aesar Supplier No. L16520.22
  • ONLINE EXCLUSIVE PRICE
    £156.06
    £127.65 / 100g
    Save £28.41
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1189-93-1 | C6H18O2Si3 | 206.463 g/mol

1,1,3,3,5,5-Hexamethyltrisiloxane is involved in the preparation of 2-bis(3-allyl-4-hydroxyphenyl)hexafluoropropane, which is associated with inorganic siloxane polymer and used as a coating material for the detection of toxic chemical warfare agents. It is also used as a silylating agent and to prepare 1,5-Bis[2-(1,2-epoxycyclohex-4-yl)ethyl]-1,1,3,3,5,5-hexamethyltrisiloxane from 4-vinyl-1-cyclohexene-1,2-epoxide by hydrosilylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,3,3,5,5-Hexamethyltrisiloxane is involved in the preparation of 2-bis(3-allyl-4-hydroxyphenyl)hexafluoropropane, which is associated with inorganic siloxane polymer and used as a coating material for the detection of toxic chemical warfare agents. It is also used as a silylating agent and to prepare 1,5-Bis[2-(1,2-epoxycyclohex-4-yl)ethyl]-1,1,3,3,5,5-hexamethyltrisiloxane from 4-vinyl-1-cyclohexene-1,2-epoxide by hydrosilylation.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189-93-1
Molecular Formula C6H18O2Si3
Molecular Weight (g/mol) 206.463
MDL Number MFCD00039788
InChI Key YTEISYFNYGDBRV-UHFFFAOYSA-N
Synonym 1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,1,3,3,5,5-hexamethyl, 1,3,3,5,5-hexamethyltrisiloxane, 2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, dimethylbis dimethylsilyloxy silane
PubChem CID 6327152
IUPAC Name [(dimethyl-$l^{3}-silanyl)oxy-dimethylsilyl]oxy-dimethylsilicon
SMILES C[Si](C)O[Si](C)(C)O[Si](C)C

Specifications

Boiling Point 128°C
Melting Point -67°C
Quantity 100 g
Flash Point 20°C (68°F)
Refractive Index 1.381
Sensitivity Moisture sensitive
Density 0.822
UN Number UN1993
Beilstein 1746341
Solubility Information Slightly miscible with water.
Formula Weight 208.48
Percent Purity 97%
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.