missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5-Hexamethyltrisiloxane, 95+%, AcroSeal™, Thermo Scientific™

Product Code. 10321390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10321390 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321390 Supplier Thermo Scientific Acros Supplier No. 433071000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 128°C
CAS 1189-93-1
Color Colorless
CAS Min % 95.0
CAS Max % 100.0
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 94% min. (GC)
Molecular Formula C6H20O2Si3
Linear Formula Si(CH3)2[OSi(CH3)2H]
Flash Point 20°C
Synonym 1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,1,3,3,5,5-hexamethyl, 1,3,3,5,5-hexamethyltrisiloxane, 2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, dimethylbis dimethylsilyloxy silane
InChI Key YTEISYFNYGDBRV-UHFFFAOYSA-N
SMILES C[Si](C)O[Si](C)(C)O[Si](C)C
IUPAC Name [(dimethyl-$l^{3}-silanyl)oxy-dimethylsilyl]oxy-dimethylsilicon
Molecular Weight (g/mol) 208.48
PubChem CID 6327152
Formula Weight 208.48
Percent Purity ≥95%
Infrared Spectrum Authentic
Packaging AcroSeal glass Bottle
Refractive Index 1.3790 to 1.382
Specific Gravity 0.822
Density 0.8220g/mL
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
Show More Show Less
compliance-icons
Product Identifier
  • 1,1,3,3,5,5-Hexamethyltrisiloxane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.