missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trichloroethane, TRC

Product Code. 30457140
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457140 5 g -
30451894 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457140 Supplier Toronto Research Chemicals Supplier No. TRCT7735305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-00-5
Molecular Formula C2 H3 Cl3
Molecular Weight (g/mol) 133.4
Synonym Ethane, 1,1,2-trichloro-, 1,1,2-Trichloroethane, 1,2,2-Trichloroethane, NSC 405074, Trichloroethane, Vinyltrichloride, β-T, β-Trichloroethane
IUPAC Name 1,1,2-trichloroethane
SMILES ClCC(Cl)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H3Cl3/c3-1-2(4)5/h2H,1H2
Formula Weight 131.93
Chemical Name or Material 1,1,2-Trichloroethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.