missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetraphenylethane-1,2-diol, TRC

Product Code. 30415373
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415373 1 g -
30421992 2.5 g -
30426313 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30415373 Supplier Toronto Research Chemicals Supplier No. TRCT3028681G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-72-2
Molecular Formula C26H22O2
Molecular Weight (g/mol) 366.45
Synonym 1,1,2,2-Tetraphenyl-1,2-ethandiol, 1,1,2,2-Tetraphenylethylene Glycol, Benzophenone Pinacol, Benzopinacone, Benzpinacol, Benzpinacone, NSC 120377, NSC 1973, Tetraphenyl-1,2-ethanediol, Tetraphenylethylene Glycol, alpha,alpha'-Bibenzhydrol
IUPAC Name 1,1,2,2-tetraphenylethane-1,2-diol
SMILES OC(c1ccccc1)(c2ccccc2)C(O)(c3ccccc3)c4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C26H22O2/c27-25(21-13-5-1-6-14-21,22-15-7-2-8-16-22)26(28,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20,27-28H
Formula Weight 366.162
Chemical Name or Material 1,1,2,2-Tetraphenylethane-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.