missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetramethylcyclopropane, 99%, Thermo Scientific Chemicals

Product Code. 11495726
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495726 5 g 5g
11405736 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495726 Supplier Thermo Scientific Alfa Aesar Supplier No. B20809.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4127-47-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00060790
InChI Key JCHUCGKEGUAHEH-UHFFFAOYSA-N
Synonym cyclopropane, 1,1,2,2-tetramethyl, acmc-209jjk, jchucgkeguaheh-uhfffaoysa, 1,1,2,2-tetramethyl cyclopropane, 1,1,2,2-tetramethyl-cyclopropane
PubChem CID 77778
IUPAC Name 1,1,2,2-tetramethylcyclopropane
SMILES CC1(C)CC1(C)C

Specifications

Density 0.72
Boiling Point 76°C
Quantity 5 g
UN Number UN3295
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material 1,1,2,2-Tetramethylcyclopropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.