missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 95%

Product Code. 11492110
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492110 5 g 5g
11482110 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492110 Supplier Thermo Scientific Alfa Aesar Supplier No. H28705.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1145-98-8 | C16H22Si2 | 270.522 g/mol

Dimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1145-98-8
Molecular Formula C16H22Si2
Molecular Weight (g/mol) 270.522
MDL Number MFCD00054786
InChI Key IIOOIYHUTINYQO-UHFFFAOYSA-N
Synonym 1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane
PubChem CID 136916
IUPAC Name [dimethyl(phenyl)silyl]-dimethyl-phenylsilane
SMILES C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2

Specifications

Boiling Point 111°CC
Melting Point 34°C to 37°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 270.52
Percent Purity 97%
Chemical Name or Material 1,1,2,2-Tetramethyl-1,2-diphenyldisilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.