missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrafluoroethyl methyl ether, 99%, Thermo Scientific™

Product Code. 11334815
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Product Code. 11334815

Brand: Thermo Scientific Alfa Aesar L17889.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 425-88-7
Molecular Formula C3H4F4O
Molecular Weight (g/mol) 132.058
MDL Number MFCD00042106
InChI Key YQQHEHMVPLLOKE-UHFFFAOYSA-N
Synonym 1,1,2,2-tetrafluoroethyl methyl ether, methyl 1,1,2,2-tetrafluoroethyl ether, methyl1,1,2,2-tetrafluoroethylether, acmc-20ap8j, ethane, 1,1,2,2-tetrafluoro-1-methoxy, hfe-254pc, methyl 2h-tetrafluoroethyl ether, yqqhehmvplloke-uhfffaoysa, 1,1,2,2-tetrafluoroethylmethylether, 1-methoxy-1,1,2,2-tetrafluoroethane
PubChem CID 67926
IUPAC Name 1,1,2,2-tetrafluoro-1-methoxyethane
SMILES COC(C(F)F)(F)F

Specifications

Melting Point -107°C
Density 1.294
Boiling Point 36°C to 37°C
Refractive Index 1.2939
Quantity 25 g
UN Number UN3271
Beilstein 1737755
Formula Weight 132.06
Percent Purity 99%
Chemical Name or Material 1,1,2,2-Tetrafluoroethyl methyl ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.