missing translation for 'onlineSavingsMsg'
Learn More

(1,1,2,2-Tetrafluoroethoxy)benzene, 98%, Thermo Scientific™

Product Code. 11480648
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11480648

Brand: Thermo Scientific Alfa Aesar B25189.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-57-2
Molecular Formula C8H6F4O
Molecular Weight (g/mol) 194.129
MDL Number MFCD00042102
InChI Key GRDIVJPQARIBNZ-UHFFFAOYSA-N
Synonym 1,1,2,2-tetrafluoroethoxy benzene, tetrafluoroethoxy benzene, fentalene-14, benzene, 1,1,2,2-tetrafluoroethoxy, 1,1,2,2-tetrafluoro-1-phenoxyethane, 3-06-00-00598 beilstein handbook reference, benzene, tetrafluoroethoxy, ether, phenyl tetrafluoroethoxy, tetrafluoroethyl ether of phenol, 4-1,1,2,2-tetrafluoroethoxy benzene
PubChem CID 67692
IUPAC Name 1,1,2,2-tetrafluoroethoxybenzene
SMILES C1=CC=C(C=C1)OC(C(F)F)(F)F

Specifications

Density 1.28
Boiling Point 152°C to 153°C
Flash Point 46°C (115°F)
Refractive Index 1.421
Quantity 1 g
UN Number UN1993
Formula Weight 194.13
Percent Purity 98%
Chemical Name or Material (1,1,2,2-Tetrafluoroethoxy)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.