missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane, 98+%

Product Code. 11331607
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331607 100 g 100g
11321607 25 g 25g
11341607 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11331607 Supplier Thermo Scientific Alfa Aesar Supplier No. L04489.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 79-34-5 | C2H2Cl4 | 167.838 g/mol

1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.

Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-34-5
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 167.838
MDL Number MFCD00000848
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
PubChem CID 6591
ChEBI CHEBI:36026
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Melting Point -43°C
Density 1.596
Boiling Point 142°C to 146°C
Linear Formula Cl2CHCHCl2
Refractive Index 1.4935
Quantity 100 g
UN Number UN1702
Beilstein 969206
Merck Index 14,9189
Solubility Information Miscible with water,methanol,ethanol,chloroform and acetone.
Formula Weight 167.85
Percent Purity ≥98%
Chemical Name or Material 1,1,2,2-Tetrachloroethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.