missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2,3,3,4,4,4-Nonafluoro-N-(2-hydroxyethyl)-N-methyl-1-butanesulfonamide, TRC

Product Code. 30011789
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011789 100 mg 100mg
30009238 1 g 1g
30003795 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011789 Supplier Toronto Research Chemicals Supplier No. TRCN649510100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34454-97-2
Molecular Formula C7 H8 F9 N O3 S
Molecular Weight (g/mol) 357.19
Synonym 2-[N-(Perfluorobutylsulfonyl)-N-methylamino]ethanol, N-(2-Hydroxyethyl)-N-methyl-perfluorobutane-1-sulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-N-(2-hydroxyethyl)-N-methyl-1-butanesulfonamide
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N-(2-hydroxyethyl)-N-methyl-butane-1-sulfonamide
SMILES CN(CCO)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H8F9NO3S/c1-17(2-3-18)21(19,20)7(15,16)5(10,11)4(8,9)6(12,13)14/h18H,2-3H2,1H3
Formula Weight 357.0081
Percent Purity >95%
Chemical Name or Material 2-(N-Methylperfluorobutanesulfamoyl)ethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.