missing translation for 'onlineSavingsMsg'
Learn More

1,1':2',1' ':2' ',1' ' '-Quaterphenyl, TRC

Product Code. 30420276
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420276 2.5 g 2.5g
30415707 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30420276 Supplier Toronto Research Chemicals Supplier No. TRCQ5094502.5G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 641-96-3
Molecular Formula C24H18
Molecular Weight (g/mol) 306.4
Synonym 2,2'-Diphenyl-biphenyl, o-Quaterphenyl, 2,2'-Diphenyl-1,1'-biphenyl, 2,2'-Diphenylbiphenyl, NSC 90717, o,o-Quaterphenyl, o,o'-Quaterphenyl
IUPAC Name 1-phenyl-2-(2-phenylphenyl)benzene
SMILES c1ccc(cc1)c2ccccc2c3ccccc3c4ccccc4

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C24H18/c1-3-11-19(12-4-1)21-15-7-9-17-23(21)24-18-10-8-16-22(24)20-13-5-2-6-14-20/h1-18H
Formula Weight 306.1409
Chemical Name or Material 1,1':2',1' ':2' ',1' ' '-Quaterphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.