missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Trifluoroacetone, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1,1,1-Trifluoroacetone |
| CAS | 421-50-1 |
| Molecular Formula | C3H3F3O |
| Quantity | 5 g |
| Synonym | 3,3,3-Trifluoroacetone, 1,1,1-Trifluoro-2-propanone, Methyl Trifluoromethyl Ketone, NSC 66412, Trifluoromethyl Methyl Ketone, alpha,alpha,alpha-Trifluoroacetone |
| InChI Formula | InChI=1S/C3H3F3O/c1-2(7)3(4,5)6/h1H3 |
| SMILES | CC(=O)C(F)(F)F |
| IUPAC Name | 1,1,1-trifluoropropan-2-one |
| Molecular Weight (g/mol) | 112.05 |
| Formula Weight | 112.0136 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction