Learn More
1,1,1-Trifluoroacetone, 98+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 421-50-1 |
---|---|
Molecular Formula | C3H3F3O |
Molecular Weight (g/mol) | 112.05 |
InChI Key | FHUDAMLDXFJHJE-UHFFFAOYSA-N |
Synonym | 1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone |
PubChem CID | 9871 |
IUPAC Name | 1,1,1-trifluoropropan-2-one |
SMILES | CC(=O)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10726161
|
Thermo Scientific Acros
139740250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
10103290
|
Thermo Scientific Acros
139741000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
11974871
|
Thermo Scientific Acros
139745000 |
500 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
421-50-1 | |
112.05 | |
1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone | |
1,1,1-trifluoropropan-2-one |
C3H3F3O | |
FHUDAMLDXFJHJE-UHFFFAOYSA-N | |
9871 | |
CC(=O)C(F)(F)F |
Specifications
421-50-1 | |
Colorless | |
22.0°C | |
Authentic | |
C3H3F3O | |
1.3000 (20°C, 589nm) max. | |
13,323; 15,212 | |
1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone | |
FHUDAMLDXFJHJE-UHFFFAOYSA-N | |
1,1,1-trifluoropropan-2-one | |
9871 | |
112.05 | |
Liquid |
-78.0°C | |
1.2500g/mL | |
−30°C | |
98% min. (GC) | |
CF3COCH3 | |
01,II,717 | |
1.25 | |
Solubility in water: miscible | |
CC(=O)C(F)(F)F | |
112.05 | |
3.28 mPa.s (15°C) | |
98+% | |
1, 1, 1-Trifluoroacetone |
Safety and Handling
GHS H Statement
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 207-005-9
- 1,1,1-Trifluoroacetone
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H224-Extremely flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only