missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-5-methyl-2,4-hexanedione, 97%, Thermo Scientific™

Product Code. 10627221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627221 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627221 Supplier Thermo Scientific Acros Supplier No. 421760250
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 30984-28-2
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Boiling Point 61°C (48.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H9F3O2
Linear Formula CF3COCH2COCH(CH3)2
Refractive Index 1.4005 to 1.4025
MDL Number MFCD00045106
Quantity 25 g
Synonym 1,1,1-trifluoro-5-methyl-2,4-hexanedione, trifluoroacetyl isobutyrylmethane, 2,4-hexanedione, 1,1,1-trifluoro-5-methyl, 5-methyl-1,1,1-trifluoro-2,4-hexanedione, 2,4-hexanedione,1,1,1-trifluoro-5-methyl, tos-bb-0654, 2,4-dioxo-5-methyl-1,1,1-trifluorohexane, 5-methyl-1,1,1-trifluorohexane-2,4-dione
InChI Key QDJWKASBKAMIDF-ZZXKWVIFSA-N
SMILES CC(C)C(=O)\C=C(\O)C(F)(F)F
IUPAC Name 1,1,1-trifluoro-5-methylhexane-2,4-dione
Molecular Weight (g/mol) 182.14
PubChem CID 160191
Formula Weight 182.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-5-methyl-2,4-hexanedione
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,1-Trifluoro-5-methyl-2,4-hexanedione
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.