missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-4-iodobutane, 98+%, stab. with copper

Product Code. 11400278
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400278 1 g 1g
11410278 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400278 Supplier Thermo Scientific Alfa Aesar Supplier No. B23811.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 461-17-6 | C4H6F3I | 237.992 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-17-6
Molecular Formula C4H6F3I
Molecular Weight (g/mol) 237.992
MDL Number MFCD00041543
InChI Key LNDGACQEAYKNOI-UHFFFAOYSA-N
Synonym 1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
PubChem CID 2775189
IUPAC Name 1,1,1-trifluoro-4-iodobutane
SMILES C(CC(F)(F)F)CI

Specifications

Density 1.85
Boiling Point 126°C to 127°C
Flash Point >100°C (212°F)
Linear Formula CF3(CH2)3I
Refractive Index 1.4335
Quantity 1 g
Beilstein 1736064
Sensitivity Light sensitive
Formula Weight 237.99
Percent Purity ≥98%
Chemical Name or Material 1,1,1-Trifluoro-4-iodobutane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.