missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-butanone, TRC

Product Code. 30456006
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456006 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456006 Supplier Toronto Research Chemicals Supplier No. TRCT3818801G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1,1-Trifluoro-2-butanone
CAS 381-88-4
Molecular Formula C4H5F3O
Quantity 1 g
Synonym Ethyl Trifluoromethyl Ketone, NSC 42602
InChI Formula InChI=1S/C4H5F3O/c1-2-3(8)4(5,6)7/h2H2,1H3
SMILES CCC(=O)C(F)(F)F
IUPAC Name 1,1,1-trifluorobutan-2-one
Molecular Weight (g/mol) 126.08
Formula Weight 126.0292
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.