missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-butanone, 96%

Product Code. 11343806
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343806 5 g 5g
11333806 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343806 Supplier Thermo Scientific Alfa Aesar Supplier No. L16868.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 381-88-4 | C4H5F3O | 126.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-88-4
Molecular Formula C4H5F3O
Molecular Weight (g/mol) 126.08
MDL Number MFCD00190641
InChI Key QBVHMPFSDVNFAY-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2-butanone, 1,1,1-trifluorobutane-2-one, 2-butanone, 1,1,1-trifluoro, 1,1,1-trifluoromethyl ethyl ketone, trifluoro-2-butanone, acmc-20ap8w, trifluoromethylethylketone, ethyl trifluoromethyl ketone, trifluoromethyl ethyl ketone, 1,1-trifluoro-2-butanone
PubChem CID 238288
IUPAC Name 1,1,1-trifluorobutan-2-one
SMILES CCC(=O)C(F)(F)F

Specifications

Density 0.929
Boiling Point 44°C to 45°C
Flash Point −17°C (1°F)
Refractive Index 1.307
Quantity 5 g
UN Number UN1224
Beilstein 1702220
Formula Weight 126.08
Percent Purity 96%
Chemical Name or Material 1,1,1-Trifluoro-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.