missing translation for 'onlineSavingsMsg'
Learn More

1-(1,1-Dimethylpropyl)-4-iodobenzene, TRC

Product Code. 30382151
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382151 100 mg 100mg
30377485 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382151 Supplier Toronto Research Chemicals Supplier No. TRCD478035100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(1,1-Dimethylpropyl)-4-iodobenzene
CAS 860556-27-0
Molecular Formula C11H15I
Quantity 100 mg
Synonym 1-tert-Amyl-4-iodo-benzene, 1-(1,1-Dimethylpropyl)-4-iodo-benzene
InChI Formula InChI=1S/C11H15I/c1-4-11(2,3)9-5-7-10(12)8-6-9/h5-8H,4H2,1-3H3
SMILES CCC(C)(C)c1ccc(I)cc1
IUPAC Name 1-iodo-4-(2-methylbutan-2-yl)benzene
Molecular Weight (g/mol) 274.14
Formula Weight 274.0218
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.