missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,5,5,5-Hexafluoroacetylacetone, 99%

Product Code. 11904881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11904881 100 g Glass bottle 100g
10051861 5 g Glass bottle 5g
10245241 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11904881 Supplier Thermo Scientific Acros Supplier No. 218531000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1522-22-1 | C5H2F6O2 | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1522-22-1
Molecular Formula C5H2F6O2
Molecular Weight (g/mol) 208.06
InChI Key QAMFBRUWYYMMGJ-UHFFFAOYSA-N
Synonym hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hfac, 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,3-bis trifluoromethyl propane-1,3-dione, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane
PubChem CID 73706
IUPAC Name 1,1,1,5,5,5-hexafluoropentane-2,4-dione
SMILES C(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.4700g/mL
Boiling Point 70.0°C to 71.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3COCH2COCF3
Refractive Index 1.3320 to 1.3340
Quantity 100 g
Beilstein 01,III,3123
Specific Gravity 1.47
Solubility Information Solubility in water: immiscible
Formula Weight 208.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1,1,5,5,5-Hexafluoroacetylacetone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1,1,5,5,5-Hexafluoroacetylacetone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.