missing translation for 'onlineSavingsMsg'
Learn More

1,1'-(1,4-phenylene)bis-Ethanone, TRC

Product Code. 30421551
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421551 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421551 Supplier Toronto Research Chemicals Supplier No. TRCP997790250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1009-61-6
Molecular Formula C10 H10 O2
Quantity 250 mg
Synonym Benzene, p-diacetyl- (6CI,8CI), 1,1'-(1,4-Phenylene)bis[ethanone], 1,1'-(1,4-Phenylene)bis(ethan-1-one), 1,1'-(1,4-Phenylene)diethanone, 1,4-Diacetylbenzene, 1-(4-Acetylphenyl)ethan-1-one, 1-(4-Acetylphenyl)ethanone, 4'-Acetylacetophenone, NSC 295548, p-Acetylacetophenone, p-Diacetylbenzene
InChI Formula InChI=1S/C10H10O2/c1-7(11)9-3-5-10(6-4-9)8(2)12/h3-6H,1-2H3
SMILES CC(=O)c1ccc(cc1)C(=O)C
IUPAC Name 1-(4-acetylphenyl)ethanone
Molecular Weight (g/mol) 162.185
Formula Weight 162.0681
Chemical Name or Material 1,4-Diacetylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.