missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3-Pentafluorobutane, 99.5+%

Product Code. 15436597
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436597 50 g 50g
15446597 250 g 250g
15426597 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 15436597 Supplier Thermo Scientific Alfa Aesar Supplier No. H33737.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 406-58-6 | C4H5F5 | 148.076 g/mol

Used as foam blowing agent in the production of closed cell rigid insulating foams. It is also used as degreasing solvent in semi-closed and closed degreasing systems. It finds also its use as heat exchange fluid in closed heat transfer systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as foam blowing agent in the production of closed cell rigid insulating foams. It is also used as degreasing solvent in semi-closed and closed degreasing systems. It finds also its use as heat exchange fluid in closed heat transfer systems.

Solubility
Soluble in water (1.7g/L at 21.2C). Miscible with most organic solvents.

Notes
Store in cool location. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 406-58-6
Molecular Formula C4H5F5
Molecular Weight (g/mol) 148.076
MDL Number MFCD03094115
InChI Key WZLFPVPRZGTCKP-UHFFFAOYSA-N
PubChem CID 67884
IUPAC Name 1,1,1,3,3-pentafluorobutane
SMILES CC(CC(F)(F)F)(F)F

Specifications

Density 1.27
Boiling Point 40°C
Flash Point −20°C (−4°F)
Linear Formula CH3CF2CH2CF3
Quantity 50 g
UN Number UN1993
Synonym hfc-365mfc, butane, 1,1,1,3,3-pentafluoro, butane,1,1,1,3,3-pentafluoro, solkane 365 mfc, 2,2,4,4,4-pentafluorobutane, 1h,1h,1h,3h,3h-perfluorobutane
Solubility Information Soluble in water (1.7g/L at 21.2C). Miscible with most organic solvents.
Formula Weight 148.08
Percent Purity ≥99.5%
Chemical Name or Material 1,1,1,3,3-Pentafluorobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.